Gene Transcripts

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
Chromosome Location
Chr. 5: 131949973 - 132012068 on Build GRCh38
UniGene ID
Hs.14945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009185.2 NP_001009185.1 21 2386
NM_001205247.1 NP_001192176.1 21 2281
NM_001205248.1 NP_001192177.1 21 2373
NM_001205250.1 NP_001192179.1 21 2383
NM_001205251.1 NP_001192180.1 20 2148
NM_015256.3 NP_056071.2 21 2386
GenBank mRNA AB020644.1 - 21 2346
AF129166.1 - 21 2316
AK122702.1 - 10 1844
AK294291.1 - 21 2256
BC026161.1 - 21 2282 4830357
BC047453.1 - 20 2070 5270879

View Full Product Details