Gene Transcripts

Gene Symbol
POC1A
Entrez Gene ID
25886
Gene Name
POC1 centriolar protein A
Gene Aliases
PIX2, SOFT, WDR51A
Chromosome Location
Chr. 3: 52075233 - 52154690 on Build GRCh38
UniGene ID
Hs.476306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161580.1 NP_001155052.1 4 751
NM_001161581.1 NP_001155053.1 4 822
NM_015426.4 NP_056241.3 4 751
XM_011533560.1 XP_011531862.1 4 473
XM_011533561.1 XP_011531863.1 4 473
XM_011533562.2 XP_011531864.1 4 473
XM_011533563.1 XP_011531865.1 4 473
XM_011533564.2 XP_011531866.1 4 473
XM_011533565.2 XP_011531867.1 4 473
XM_017006104.1 XP_016861593.1 4 544
GenBank mRNA AK075289.1 - 4 466
AK315688.1 - 4 473
BC007417.2 - 3 401
BC110877.1 - 4 751 5588109
BC119692.2 - 4 461 40116721
BC119693.2 - 4 537

View Full Product Details