Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308103.1 | NP_001295032.1 | 5 | 1342 | |
NM_022073.3 | NP_071356.1 | 5 | 1876 | ||
GenBank mRNA | AJ310545.1 | - | 5 | 1876 | |
AK025273.1 | - | 5 | 1876 | ||
AK026918.1 | - | 2 | 914 | ||
AK123350.1 | - | 6 | 1705 | ||
BC111057.1 | - | 5 | 1859 | 5747720 |