Gene Transcripts
- Gene Symbol
- SECISBP2
- Entrez Gene ID
- 79048
- Gene Name
- SECIS binding protein 2
- Gene Aliases
- SBP2
- Chromosome Location
- Chr. 9: 89318473 - 89367117 on Build GRCh38
- UniGene ID
- Hs.59804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282688.1 |
NP_001269617.1
|
5, 6 |
910 |
|
NM_001282689.1 |
NP_001269618.1
|
5, 6 |
792 |
|
NM_001282690.1 |
NP_001269619.1
|
5, 6 |
990 |
|
NM_024077.4 |
NP_076982.3
|
5, 6 |
913 |
|
XM_005252196.2 |
XP_005252253.1
|
5, 6 |
880 |
|
XM_006717282.2 |
XP_006717345.1
|
5, 6 |
846 |
|
XM_011519000.2 |
XP_011517302.1
|
5, 6 |
967 |
|
XM_011519001.1 |
XP_011517303.1
|
5, 6 |
967 |
|
XM_011519002.1 |
XP_011517304.1
|
5, 6 |
967 |
|
XM_011519003.1 |
XP_011517305.1
|
5, 6 |
967 |
|
XM_017015122.1 |
XP_016870611.1
|
5, 6 |
880 |
|
XM_017015123.1 |
XP_016870612.1
|
5, 6 |
880 |
|
XM_017015124.1 |
XP_016870613.1
|
5, 6 |
880 |
|
XM_017015125.1 |
XP_016870614.1
|
3, 4 |
506 |
|
GenBank mRNA |
AB208940.1 |
- |
5, 6 |
871 |
|
AF380995.1 |
- |
5, 6 |
866 |
|
AK090608.1 |
- |
5, 6 |
880 |
|
AK290182.1 |
- |
5, 6 |
759 |
|
AK299161.1 |
- |
5, 6 |
1001 |
|
AK302852.1 |
- |
5, 6 |
908 |
|
AL136881.1 |
- |
5, 6 |
746 |
|
BC001189.1 |
- |
5, 6 |
746 |
|
BC036109.1 |
- |
5, 6 |
875 |
5296292 |
BC068466.1 |
- |
5, 6 |
848 |
|
BC099900.1 |
- |
5, 6 |
849 |
|
View Full Product Details