Gene Transcripts
- Gene Symbol
- STAG2
- Entrez Gene ID
- 10735
- Gene Name
- stromal antigen 2
- Gene Aliases
- SA-2, SA2, SCC3B, bA517O1.1
- Chromosome Location
- Chr. X: 123960560 - 124102656 on Build GRCh38
- UniGene ID
- Hs.496710
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042749.2 |
NP_001036214.1
|
12, 13 |
1593 |
|
NM_001042750.1 |
NP_001036215.1
|
12, 13 |
1437 |
|
NM_001042751.1 |
NP_001036216.1
|
12, 13 |
1437 |
|
NM_001282418.1 |
NP_001269347.1
|
12, 13 |
1416 |
|
NM_006603.4 |
NP_006594.3
|
11, 12 |
1372 |
|
XM_005262357.2 |
XP_005262414.1
|
11, 12 |
1462 |
|
XM_005262358.2 |
XP_005262415.1
|
12, 13 |
1399 |
|
XM_005262359.3 |
XP_005262416.1
|
11, 12 |
1352 |
|
XM_005262360.2 |
XP_005262417.1
|
11, 12 |
1281 |
|
XM_005262361.2 |
XP_005262418.1
|
11, 12 |
1409 |
|
XM_006724727.1 |
XP_006724790.1
|
13, 14 |
1541 |
|
XM_006724728.3 |
XP_006724791.1
|
12, 13 |
1475 |
|
XM_011531253.2 |
XP_011529555.1
|
11, 12 |
1334 |
|
XM_017029232.1 |
XP_016884721.1
|
12, 13 |
1399 |
|
XM_017029233.1 |
XP_016884722.1
|
11, 12 |
1281 |
|
XM_017029234.1 |
XP_016884723.1
|
11, 12 |
1334 |
|
XM_017029235.1 |
XP_016884724.1
|
12, 13 |
1480 |
|
GenBank mRNA |
AK098737.1 |
- |
11, 12 |
1416 |
|
AK313559.1 |
- |
11, 12 |
1264 |
|
AL831939.2 |
- |
11, 12 |
1340 |
|
BC001765.1 |
- |
11, 12 |
1321 |
3352347 |
BX640970.1 |
- |
12, 13 |
1448 |
|
BX641002.1 |
- |
11, 12 |
1341 |
|
BX641003.1 |
- |
12, 13 |
1406 |
|
CR627425.1 |
- |
12, 13 |
1415 |
|
CR749433.1 |
- |
12, 13 |
1406 |
|
View Full Product Details