Gene Transcripts

Gene Symbol
DEDD
Entrez Gene ID
9191
Gene Name
death effector domain containing
Gene Aliases
CASP8IP1, DEDD1, DEFT, FLDED1, KE05
Chromosome Location
Chr. 1: 161120977 - 161132783 on Build GRCh38
UniGene ID
Hs.744092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039711.1 NP_001034800.1 2 403
NM_001039712.1 NP_001034801.1 3 374
NM_032998.2 NP_127491.1 3 436
XM_005245597.3 XP_005245654.1 3 347
XM_005245598.4 XP_005245655.1 3 1486
XM_005245599.2 XP_005245656.1 3 390
XM_005245600.2 XP_005245657.1 2 498
XM_017002802.1 XP_016858291.1 3 1486
GenBank mRNA AF043733.1 - 1 251
AF064605.3 - 2 288
AF083236.1 - 1 201
AF100341.1 - 1 201
AJ010973.1 - 1 201
AK001497.1 - 3 339
AK022531.1 - 3 324
BC013910.2 - 3 416 3504847
BC016724.1 - 3 337 4077893
BG565440.1 - 3 375
CR536556.1 - 1 201

View Full Product Details