Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286445.1 | NP_001273374.1 | 3 | 459 | |
NM_001286446.1 | NP_001273375.1 | 3 | 562 | ||
NM_001286447.1 | NP_001273376.1 | 3 | 659 | ||
NM_014722.3 | NP_055537.2 | 3 | 597 | ||
NM_015864.3 | NP_056948.2 | 3 | 659 | ||
XM_006715275.2 | XP_006715338.1 | 3 | 676 | ||
XM_006715277.1 | XP_006715340.1 | 3 | 597 | ||
XM_006715279.2 | XP_006715342.1 | 3 | 654 | ||
XM_006715281.3 | XP_006715344.1 | 3 | 676 | ||
XM_011515007.1 | XP_011513309.1 | 4 | 1185 | ||
XM_011515008.2 | XP_011513310.1 | 4 | 516 | ||
XM_011515009.1 | XP_011513311.1 | 3 | 388 | ||
XM_011515010.2 | XP_011513312.1 | 3 | 429 | ||
XM_011515012.1 | XP_011513314.1 | 3 | 676 | ||
XM_017011523.1 | XP_016867012.1 | 3 | 351 | ||
XM_017011524.1 | XP_016867013.1 | 3 | 605 | ||
XM_017011525.1 | XP_016867014.1 | 3 | 597 | ||
XM_017011526.1 | XP_016867015.1 | 3 | 597 | ||
XM_017011527.1 | XP_016867016.1 | 3 | 429 | ||
GenBank mRNA | AB002384.1 | - | 3 | 395 | |
AK299047.1 | - | 3 | 367 | ||
AK300138.1 | - | 3 | 607 | ||
AK300950.1 | - | 3 | 412 | ||
BC001232.1 | - | 3 | 649 | 3453890 | |
CN313643.1 | - | 3 | 659 | ||
DA258515.1 | - | 3 | 461 | ||
DC406827.1 | - | 3 | 563 | ||
DC428807.1 | - | 3 | 459 | ||
U49187.1 | - | 3 | 520 |