Gene Transcripts

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
Chromosome Location
Chr. 7: 134985169 - 135135782 on Build GRCh38
UniGene ID
Hs.648616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178563.3 NP_848658.3 4, 5 564
XM_011516122.2 XP_011514424.1 3, 4 370
XM_017012136.1 XP_016867625.1 4, 5 1886
XM_017012137.1 XP_016867626.1 4, 5 2168
XM_017012138.1 XP_016867627.1 3, 4 370
XM_017012139.1 XP_016867628.1 3, 4 370
XM_017012140.1 XP_016867629.1 3, 4 370
XM_017012142.1 XP_016867631.1 3, 4 370
XM_017012143.1 XP_016867632.1 3, 4 370
GenBank mRNA AK302827.1 - 4, 5 564
BC030651.2 - 4, 5 539 5267615

View Full Product Details