Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177350.5 | NP_796324.1 | 5 | 709 | |
GenBank mRNA | AF548022.1 | - | 5 | 365 | |
AK031523.1 | - | 5 | 710 | ||
BC125025.1 | - | 5 | 689 | 40058551 | |
BC125026.1 | - | 5 | 689 | 40058552 |