Gene Transcripts
- Gene Symbol
- Jag2
- Entrez Gene ID
- 16450
- Gene Name
- jagged 2
- Gene Aliases
- D12Ggc2e, Serh, mJagged2-1, sm
- Chromosome Location
- Chr. 12: 112907803 - 112929764 on Build GRCm38
- UniGene ID
- Mm.186146
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010588.2 |
NP_034718.2
|
21, 22 |
2608 |
|
GenBank mRNA |
AF038572.1 |
- |
21, 22 |
2608 |
|
View Full Product Details