Gene Transcripts

Gene Symbol
PDIA3
Entrez Gene ID
2923
Gene Name
protein disulfide isomerase family A member 3
Gene Aliases
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
Chromosome Location
Chr. 15: 43746392 - 43772606 on Build GRCh38
UniGene ID
Hs.591095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005313.4 NP_005304.3 7 930
GenBank mRNA AK075455.1 - 8 1044
AK075481.1 - 8 831
AK293249.1 - 6 603
BC014433.1 - 7 871 3542405
BC036000.1 - 7 884 4712175
BC071878.1 - 7 873 6380973
D16234.1 - 7 849
D83485.1 - 7 804
EU668338.1 - 7 884
FJ224330.1 - 7 873
U42068.1 - 7 860
Z49835.1 - 7 888

Gene Symbol
PDIA3P1
Entrez Gene ID
171423
Gene Name
protein disulfide isomerase family A member 3 pseudogene 1
Gene Aliases
ERp60, GRP58P, PDIA3P
Chromosome Location
Chr. 1: 147177851 - 147179949 on Build GRCh38
UniGene ID
Hs.658566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002305.1 1 1045
GenBank mRNA AK295985.1 - 1 930

View Full Product Details