Gene Transcripts

Gene Symbol
Bmper
Entrez Gene ID
73230
Gene Name
BMP-binding endothelial regulator
Gene Aliases
3110056H04Rik, CV-2, Crim3, Cv2
Chromosome Location
Chr. 9: 23223076 - 23485215 on Build GRCm38
UniGene ID
Mm.335020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028472.2 NP_082748.1 5 999
GenBank mRNA AF454954.1 - 5 999
AK014221.1 - 3 205
AY263358.1 - 5 520
BC066153.1 - 5 504 30098453

View Full Product Details