Gene Transcripts

Gene Symbol
Kctd15
Entrez Gene ID
233107
Gene Name
potassium channel tetramerisation domain containing 15
Gene Aliases
BC031749
Chromosome Location
Chr. 7: 34639015 - 34655507 on Build GRCm38
UniGene ID
Mm.486198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146188.1 NP_666300.1 6 2102
GenBank mRNA AK034731.1 - 7 2174
BC031749.1 - 6 2102 4507203

View Full Product Details