Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_182819.2 | NP_877971.2 | 3 | 239 | |
GenBank mRNA | AY339885.1 | - | 3 | 199 | |
BC079123.1 | - | 3 | 239 | 7126762 | |
FQ210062.1 | - | 3 | 234 | ||
FQ213156.1 | - | 3 | 270 | ||
FQ214913.1 | - | 3 | 269 | ||
FQ217174.1 | - | 3 | 264 | ||
FQ217253.1 | - | 3 | 257 | ||
FQ217570.1 | - | 3 | 270 | ||
FQ217933.1 | - | 3 | 247 | ||
FQ217995.1 | - | 3 | 269 | ||
FQ218015.1 | - | 3 | 269 | ||
FQ218434.1 | - | 3 | 269 | ||
FQ221598.1 | - | 3 | 269 | ||
FQ221667.1 | - | 3 | 268 | ||
FQ222019.1 | - | 3 | 268 | ||
FQ222086.1 | - | 3 | 257 | ||
FQ222256.1 | - | 3 | 269 | ||
FQ222576.1 | - | 3 | 261 | ||
FQ223858.1 | - | 3 | 257 | ||
FQ224255.1 | - | 3 | 257 | ||
FQ224424.1 | - | 3 | 269 | ||
FQ224448.1 | - | 3 | 269 | ||
FQ225820.1 | - | 3 | 269 | ||
FQ229714.1 | - | 3 | 270 |