Gene Transcripts

Gene Symbol
Npvf
Entrez Gene ID
60570
Gene Name
neuropeptide VF precursor
Gene Aliases
Rfrp
Chromosome Location
Chr. 4: 80391785 - 80395476 on Build Rnor_6.0
UniGene ID
Rn.62354
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023952.1 NP_076442.1 1 118
GenBank mRNA AB040288.1 - 1 118
AF330059.2 - 1 118

View Full Product Details