Gene Transcripts
- Gene Symbol
- TMEM9
- Entrez Gene ID
- 252839
- Gene Name
- transmembrane protein 9
- Gene Aliases
- DERM4, TMEM9A
- Chromosome Location
- Chr. 1: 201134771 - 201171625 on Build GRCh38
- UniGene ID
- Hs.181444
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288564.1 |
NP_001275493.1
|
4, 5 |
404 |
|
NM_001288565.1 |
NP_001275494.1
|
3, 4 |
907 |
|
NM_001288566.1 |
NP_001275495.1
|
4, 5 |
505 |
|
NM_001288567.1 |
NP_001275496.1
|
4, 5 |
719 |
|
NM_001288568.1 |
NP_001275497.1
|
4, 5 |
557 |
|
NM_001288569.1 |
NP_001275498.1
|
4, 5 |
418 |
|
NM_001288570.1 |
NP_001275499.1
|
4, 5 |
431 |
|
NM_001288571.1 |
NP_001275500.1
|
4, 5 |
497 |
|
NM_016456.4 |
NP_057540.1
|
4, 5 |
422 |
|
XM_011509385.1 |
XP_011507687.1
|
4, 5 |
481 |
|
XM_011509386.2 |
XP_011507688.1
|
4, 5 |
456 |
|
XM_011509387.2 |
XP_011507689.1
|
4, 5 |
344 |
|
GenBank mRNA |
AB013909.1 |
- |
3, 4 |
275 |
|
AF151020.1 |
- |
5, 6 |
394 |
|
AK054883.1 |
- |
4, 5 |
377 |
|
AK075335.1 |
- |
4, 5 |
332 |
|
AK091726.1 |
- |
3, 4 |
792 |
|
AK303843.1 |
- |
4, 5 |
443 |
|
AL136600.1 |
- |
4, 5 |
361 |
|
AL516746.3 |
- |
4, 5 |
596 |
|
AY138587.1 |
- |
4, 5 |
333 |
|
AY359012.1 |
- |
4, 5 |
396 |
|
BC001106.2 |
- |
4, 5 |
346 |
3506442 |
BE782946.1 |
- |
4, 5 |
383 |
|
CR457162.1 |
- |
3, 4 |
275 |
|
CR533467.1 |
- |
3, 4 |
281 |
|
DA141558.1 |
- |
4, 5 |
404 |
|
DA364005.1 |
- |
4, 5 |
422 |
|
DB031447.1 |
- |
4, 5 |
398 |
|
View Full Product Details