Gene Transcripts
- Gene Symbol
- INTS4P1
- Entrez Gene ID
- 285905
- Gene Name
- integrator complex subunit 4 pseudogene 1
- Gene Aliases
- INTS4L1
- Chromosome Location
- Chr. 7: 65141037 - 65197161 on Build GRCh38
- UniGene ID
- Hs.567827
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057766.1 |
- |
14 |
2157 |
|
View Full Product Details