Gene Transcripts

Gene Symbol
TET3
Entrez Gene ID
200424
Gene Name
tet methylcytosine dioxygenase 3
Gene Aliases
hCG_40738
Chromosome Location
Chr. 2: 73982036 - 74108176 on Build GRCh38
UniGene ID
Hs.516107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005264187.3 XP_005264244.1 2 151
XM_011532682.2 XP_011530984.1 2 151
XM_011532683.2 XP_011530985.1 2 460
XM_011532684.2 XP_011530986.1 2 1892
XM_011532685.2 XP_011530987.1 2 151
XM_011532686.2 XP_011530988.1 2 151
XM_011532688.2 XP_011530990.1 2 151
XM_017003566.1 XP_016859055.1 2 1991

View Full Product Details