Gene Transcripts
- Gene Symbol
- FRMD4B
- Entrez Gene ID
- 23150
- Gene Name
- FERM domain containing 4B
- Gene Aliases
- 6030440G05Rik, GRSP1
- Chromosome Location
- Chr. 3: 69168782 - 69541340 on Build GRCh38
- UniGene ID
- Hs.371681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015123.2 |
NP_055938.1
|
19, 20 |
2233 |
|
XM_005264722.1 |
XP_005264779.1
|
19, 20 |
1910 |
|
XM_005264723.1 |
XP_005264780.1
|
9, 10 |
1392 |
|
XM_017005989.1 |
XP_016861478.1
|
19, 20 |
2017 |
|
XM_017005990.1 |
XP_016861479.1
|
19, 20 |
1910 |
|
XM_017005991.1 |
XP_016861480.1
|
20, 21 |
2070 |
|
XM_017005992.1 |
XP_016861481.1
|
19, 20 |
1850 |
|
XM_017005993.1 |
XP_016861482.1
|
19, 20 |
1841 |
|
XM_017005994.1 |
XP_016861483.1
|
19, 20 |
3683 |
|
XM_017005995.1 |
XP_016861484.1
|
13, 14 |
1427 |
|
XM_017005996.1 |
XP_016861485.1
|
9, 10 |
1129 |
|
XM_017005997.1 |
XP_016861486.1
|
9, 10 |
1392 |
|
GenBank mRNA |
AB023230.1 |
- |
19, 20 |
2002 |
|
AK024086.1 |
- |
3, 4 |
268 |
|
AK295045.1 |
- |
14, 15 |
1475 |
|
AL832231.1 |
- |
9, 10 |
1230 |
|
BC110637.1 |
- |
4, 5 |
454 |
|
View Full Product Details