Gene Transcripts

Gene Symbol
TTTY23
Entrez Gene ID
252955
Gene Name
testis-specific transcript, Y-linked 23 (non-protein coding)
Gene Aliases
NCRNA00148, TTTY23B
Chromosome Location
Chr. Y: 9910798 - 9911962 on Build GRCh38
UniGene ID
Hs.522846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001540.1 1, 2 66

Gene Symbol
TTTY23B
Entrez Gene ID
100101121
Gene Name
testis-specific transcript, Y-linked 23B (non-protein coding)
Gene Aliases
TTTY23
Chromosome Location
Chr. Y: 6242446 - 6243610 on Build GRCh38
UniGene ID
Hs.522846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003593.1 1, 2 66

View Full Product Details