Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022353.2 | NP_071748.2 | 3 | 989 | |
XM_005246766.3 | XP_005246823.1 | 3 | 589 | ||
XM_006712685.1 | XP_006712748.1 | 3 | 701 | ||
XM_006712686.1 | XP_006712749.1 | 3 | 572 | ||
XM_011511631.1 | XP_011509933.1 | 3 | 720 | ||
XM_017004676.1 | XP_016860165.1 | 3 | 589 | ||
XM_017004677.1 | XP_016860166.1 | 3 | 589 | ||
XM_017004678.1 | XP_016860167.1 | 3 | 701 | ||
XM_017004679.1 | XP_016860168.1 | 3 | 720 | ||
XM_017004680.1 | XP_016860169.1 | 3 | 675 | ||
XM_017004681.1 | XP_016860170.1 | 3 | 544 | ||
XM_017004682.1 | XP_016860171.1 | 3 | 572 | ||
XM_017004683.1 | XP_016860172.1 | 3 | 572 | ||
XM_017004684.1 | XP_016860173.1 | 3 | 481 | ||
XM_017004685.1 | XP_016860174.1 | 3 | 710 | ||
GenBank mRNA | AJ295148.1 | - | 2 | 478 | |
AK055441.1 | - | 4 | 798 | ||
AK294841.1 | - | 3 | 586 | ||
AK314565.1 | - | 3 | 989 | ||
BC011904.2 | - | 3 | 558 | 4121450 |