Gene Transcripts

Gene Symbol
C12orf49
Entrez Gene ID
79794
Gene Name
chromosome 12 open reading frame 49
Gene Aliases
Chromosome Location
Chr. 12: 116713320 - 116738070 on Build GRCh38
UniGene ID
Hs.592011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024738.2 NP_079014.1 2 297
GenBank mRNA AK025068.1 - 2 265
AK222983.1 - 2 277
BC019843.1 - 2 157 5112874

View Full Product Details