Gene Transcripts

Gene Symbol
TEX13A
Entrez Gene ID
56157
Gene Name
testis expressed 13A
Gene Aliases
Chromosome Location
Chr. X: 105218929 - 105220693 on Build GRCh38
UniGene ID
Hs.567543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291277.1 NP_001278206.1 2 527
NM_031274.4 NP_112564.1 2 553
GenBank mRNA AF285597.1 - 2 534
BC042547.1 - 2 508 5164310
BC108734.1 - 2 526 5744723

View Full Product Details