Gene Transcripts
- Gene Symbol
- Tmbim6
- Entrez Gene ID
- 24822
- Gene Name
- transmembrane BAX inhibitor motif containing 6
- Gene Aliases
- Ab1-011, Ac1-149, Cc1-27, Tegt
- Chromosome Location
- Chr. 7: 141034980 - 141070269 on Build Rnor_6.0
- UniGene ID
- Rn.107678
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019381.2 |
NP_062254.2
|
6, 7 |
523 |
|
XM_006257323.2 |
XP_006257385.1
|
6, 7 |
665 |
|
XM_008765670.1 |
XP_008763892.1
|
7, 8 |
623 |
|
GenBank mRNA |
AY321320.1 |
- |
8, 9 |
661 |
|
AY325130.1 |
- |
8, 9 |
661 |
|
AY325243.1 |
- |
8, 9 |
661 |
|
BC058478.1 |
- |
6, 7 |
529 |
6888036 |
FQ217052.1 |
- |
6, 7 |
522 |
|
FQ218664.1 |
- |
6, 7 |
525 |
|
FQ226858.1 |
- |
6, 7 |
552 |
|
FQ229652.1 |
- |
6, 7 |
524 |
|
FQ230608.1 |
- |
6, 7 |
552 |
|
FQ231817.1 |
- |
6, 7 |
530 |
|
X75855.1 |
- |
6, 7 |
552 |
|
X75856.1 |
- |
5, 6 |
436 |
|
View Full Product Details