Gene Transcripts
- Gene Symbol
- Lenep
- Entrez Gene ID
- 113917
- Gene Name
- lens epithelial protein
- Gene Aliases
- Elp, Lep503
- Chromosome Location
- Chr. 2: 188727004 - 188727670 on Build Rnor_6.0
- UniGene ID
- Rn.10187
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053614.1 |
NP_446066.1
|
1 |
208 |
|
GenBank mRNA |
U15149.1 |
- |
1 |
208 |
|
- Gene Symbol
- Flad1
- Entrez Gene ID
- 751787
- Gene Name
- flavin adenine dinucleotide synthetase 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 188726994 - 188736462 on Build Rnor_6.0
- UniGene ID
- Rn.191905
- Species
- Rattus norvegicus
View Full Product Details