Gene Transcripts

Gene Symbol
RXFP1
Entrez Gene ID
59350
Gene Name
relaxin/insulin like family peptide receptor 1
Gene Aliases
LGR7, RXFPR1
Chromosome Location
Chr. 4: 158521711 - 158653372 on Build GRCh38
UniGene ID
Hs.591686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253727.1 NP_001240656.1 3 538
NM_001253729.1 NP_001240658.1 3 538
NM_001253732.1 NP_001240661.1 3 538
NM_001253733.1 NP_001240662.1 3 538
NM_021634.3 NP_067647.2 3 538
NR_045579.1 4 1227
NR_045580.1 3 591
NR_045581.1 3 562
NR_045582.1 3 562
NR_045583.1 2 400
NR_045584.1 3 538
XM_011532174.1 XP_011530476.1 3 507
XM_011532175.1 XP_011530477.1 3 507
XM_011532176.1 XP_011530478.1 3 510
XM_011532179.2 XP_011530481.1 3 507
XM_017008517.1 XP_016864006.1 3 507
XM_017008518.1 XP_016864007.1 3 510
XM_017008519.1 XP_016864008.1 4 471
XM_017008520.1 XP_016864009.1 3 560
XM_017008521.1 XP_016864010.1 4 471
XM_017008522.1 XP_016864011.1 3 559
XM_017008523.1 XP_016864012.1 3 510
XM_017008524.1 XP_016864013.1 3 510
GenBank mRNA AB208943.1 - 3 276
AF190500.1 - 3 275
AK122647.1 - 5 579
AK294694.1 - 3 302
AK295040.1 - 4 1050
AK307889.1 - 2 299
AK312470.1 - 3 370
AY899848.1 - 3 275
AY899849.1 - 3 275
BC112142.1 - 3 357 8327610
BC113617.1 - 3 360 8322669
BC143556.1 - 3 380
BC143558.1 - 3 413
BX647980.1 - 3 476
BX647985.1 - 3 505
DA241507.1 - 3 511

View Full Product Details