Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010922.2 | NP_035052.2 | 3 | 317 | |
GenBank mRNA | AA240860.1 | - | 3 | 314 | |
AF034092.1 | - | 3 | 317 | ||
AK018757.1 | - | 2 | 83 | ||
AK145885.1 | - | 3 | 315 | ||
BC028436.1 | - | 3 | 282 | 1246513 |