Gene Transcripts
- Gene Symbol
- Jam2
- Entrez Gene ID
- 67374
- Gene Name
- junction adhesion molecule 2
- Gene Aliases
- 1110002N23Rik, 2410030G21Rik, 2410167M24Rik, JAM-2, JAM-B, Jcam2, VE-JAM
- Chromosome Location
- Chr. 16: 84774123 - 84826380 on Build GRCm38
- UniGene ID
- Mm.41758
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023844.5 |
NP_076333.3
|
2, 3 |
510 |
|
GenBank mRNA |
AF255911.1 |
- |
2, 3 |
233 |
|
AJ291757.1 |
- |
2, 3 |
446 |
|
AK010616.1 |
- |
2, 3 |
352 |
|
AK010826.1 |
- |
2, 3 |
354 |
|
AK013914.1 |
- |
2, 3 |
351 |
|
AK028757.1 |
- |
2, 3 |
201 |
|
AK045095.1 |
- |
2, 3 |
248 |
|
AK078128.1 |
- |
2, 3 |
236 |
|
AK153636.1 |
- |
2, 3 |
200 |
|
BC028778.1 |
- |
2, 3 |
510 |
1195543 |
View Full Product Details