Gene Transcripts
- Gene Symbol
- Dph1
- Entrez Gene ID
- 116905
- Gene Name
- DPH1 homolog (S. cerevisiae)
- Gene Aliases
- 2310011M22Rik, 4930488F09Rik, AW551873, Dph2l1, Ovca1
- Chromosome Location
- Chr. 11: 75177643 - 75190483 on Build GRCm38
- UniGene ID
- Mm.41496
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082812.1 |
- |
13 |
2114 |
|
AK128996.1 |
- |
|
2100 |
|
- Gene Symbol
- Ovca2
- Entrez Gene ID
- 246257
- Gene Name
- candidate tumor suppressor in ovarian cancer 2
- Gene Aliases
- 9030414M07Rik
- Chromosome Location
- Chr. 11: 75175943 - 75178808 on Build GRCm38
- UniGene ID
- Mm.41496
- Species
- Mus musculus
View Full Product Details