Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144491.2 | NP_652762.2 | 2, 3 | 245 | |
GenBank mRNA | AK082812.1 | - | 2, 3 | 250 | |
AK128996.1 | - | 236 | |||
AK219998.1 | - | 1, 2 | 165 | ||
AY078170.1 | - | 2, 3 | 236 | ||
BB651982.1 | - | 2, 3 | 250 | ||
BC044663.1 | - | 2, 3 | 216 | ||
BC080761.1 | - | 2, 3 | 221 |