Gene Transcripts
- Gene Symbol
- Frmd4b
- Entrez Gene ID
- 232288
- Gene Name
- FERM domain containing 4B
- Gene Aliases
- 6030440G05Rik, C87375, GOBLIN, GRSP1, R74720, mKIAA1013
- Chromosome Location
- Chr. 6: 97286867 - 97617657 on Build GRCm38
- UniGene ID
- Mm.27789
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145148.2 |
NP_660130.2
|
5, 6 |
852 |
|
GenBank mRNA |
AB042027.1 |
- |
5, 6 |
775 |
|
AF327856.1 |
- |
4, 5 |
475 |
|
AF327857.1 |
- |
3, 4 |
262 |
|
AK051779.1 |
- |
5, 6 |
724 |
|
AK129262.1 |
- |
4, 5 |
327 |
|
AK132280.1 |
- |
5, 6 |
716 |
|
AK143978.1 |
- |
4, 5 |
474 |
|
AK160775.1 |
- |
1, 2 |
288 |
|
BC052390.1 |
- |
5, 6 |
721 |
5718942 |
View Full Product Details