Gene Transcripts

Gene Symbol
Ank3
Entrez Gene ID
11735
Gene Name
ankyrin 3, epithelial
Gene Aliases
2900054D09Rik, AI314020, Ank-3, AnkG, Ankyrin-3, Ankyrin-G
Chromosome Location
Chr. 10: 69533580 - 70027436 on Build GRCm38
UniGene ID
Mm.235960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009670.4 NP_033800.2 5 1169
NM_146005.3 NP_666117.2 28 3640
NM_170687.3 NP_733788.2 5 1169
NM_170688.2 NP_733789.1 28 3640
NM_170689.2 NP_733790.2 27 3586
NM_170690.2 NP_733791.2 27 3577
NM_170728.2 NP_733924.2 26 3523
NM_170729.2 NP_733925.2 26 3523
NM_170730.2 NP_733926.2 3 859
GenBank mRNA AF212924.1 - 3 359
AK035681.1 - 5 897
AK141720.1 - 28 3495
AK157735.1 - 6 917
AK162007.1 - 5 827
BC021657.1 - 26 3393 4188590
BC026152.1 - 5 770
L40631.1 - 5 1172
L40632.1 - 28 3182
U89274.1 - 7 786
U89275.1 - 7 786

View Full Product Details