Gene Transcripts

Gene Symbol
Dzank1
Entrez Gene ID
241688
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
2810039F03Rik, 6330439K17Rik, 6430408C01, AI851115, Ankrd64
Chromosome Location
Chr. 2: 144470555 - 144527425 on Build GRCm38
UniGene ID
Mm.442156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172859.2 NP_766447.2 4 429
GenBank mRNA AK032182.1 - 4 429
AK032632.1 - 4 420
AK163994.1 - 4 420
BC089524.1 - 4 396
BC141156.1 - 4 328 9055768
BC145464.1 - 4 328 9054026

View Full Product Details