Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173180.3 | NP_775272.1 | 2 | 208 | |
GenBank mRNA | AK004549.1 | - | 2 | 208 | |
AK032081.1 | - | 2 | 208 | ||
AK075965.1 | - | 2 | 209 | ||
AK077218.1 | - | 2 | 209 | ||
AK147110.1 | - | 2 | 211 | ||
AK150118.1 | - | 2 | 209 | ||
AK158990.1 | - | 2 | 176 | ||
AK161291.1 | - | 2 | 209 | ||
AK167975.1 | - | 2 | 209 | ||
AK169325.1 | - | 2 | 203 | ||
AK169342.1 | - | 2 | 203 | ||
BC010810.1 | - | 2 | 193 | 4241697 |