Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001383.3 | NP_001374.3 | 6, 7 | 713 | |
GenBank mRNA | AF321876.1 | - | 6, 7 | 713 | |
AK297947.1 | - | 5, 6 | 731 | ||
AK314015.1 | - | 6, 7 | 699 | ||
BC003099.1 | - | 6, 7 | 675 | ||
BC096088.1 | - | 5, 6 | 569 | 40000799 | |
BT019878.1 | - | 4, 5 | 455 | ||
S81752.1 | - | 6, 7 | 748 | ||
U34880.1 | - | 6, 7 | 748 |