Gene Transcripts

Gene Symbol
Agbl3
Entrez Gene ID
76223
Gene Name
ATP/GTP binding protein-like 3
Gene Aliases
2900053G10Rik, 4930431N21Rik, 6530406M24Rik, CCP3
Chromosome Location
Chr. 6: 34780432 - 34862184 on Build GRCm38
UniGene ID
Mm.150047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289656.1 NP_001276585.1 7 1228
NM_001289657.1 NP_001276586.1 6 1213
NM_178630.4 NP_848745.2 6 1213
GenBank mRNA AK018326.1 - 7 1057
AK029793.1 - 7 1229
AK029798.1 - 6 797
AK030378.1 - 6 771
AK047649.1 - 6 770
AK054066.1 - 5 2244
BC139065.1 - 6 665 8862087
DQ867031.1 - 6 1214
DQ867032.1 - 6 1214

View Full Product Details