Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE65
Entrez Gene ID
814656
Gene Name
Cyclin/Brf1-like TBP-binding protein
Gene Aliases
AT2G01280, F10A8.16, F10A8_16, maternal effect embryo arrest 65
Chromosome Location
Chr.2: 145676 - 149394 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335052.1 NP_001325195.1 9 - 10 681 114
NM_001335053.1 NP_001325196.1 9 - 10 681 114
NM_126189.4 NP_178237.3 9 - 10 681 114

View Full Product Details