Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MS5
- Entrez Gene ID
-
827838
- Gene Name
- Tetratricopeptide repeat (TPR)-like superfamily protein
- Gene Aliases
-
AT4G20900, MALE-STERILE 5, T13K14.60, T13K14_60, TDM1
- Chromosome Location
-
Chr.4: 11184103 - 11185844 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118208.1 |
NP_193822.1 |
4 - 5 |
693 |
114 |
View Full Product Details