Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AT2G16730
- Entrez Gene ID
-
816174
- Gene Name
- glycosyl hydrolase family 35 protein
- Gene Aliases
-
AT2G16730, BGAL13, T24I21.14, T24I21_14, beta-galactosidase 13
- Chromosome Location
-
Chr.2: 7261783 - 7266346 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127225.3 |
NP_179264.2 |
14 - 15 |
1942 |
84 |
View Full Product Details