Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCI20
Entrez Gene ID
831709
Gene Name
non-intrinsic ABC protein 9
Gene Aliases
ARABIDOPSIS THALIANA NON-INTRINSIC ABC PROTEIN 9, AT5G02270, ATNAP9, ATP-binding cassette I20, NAP9, T1E22.30, T1E22_30, non-intrinsic ABC protein 9
Chromosome Location
Chr.5: 467038 - 469272 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203276.1 NP_001190205.1 5 - 6 669 94
NM_120305.3 NP_195847.1 4 - 5 753 94
GenBank mRNA AF370554.1 - 4 - 5 533 94
AY086132.1 - 4 - 5 561 94
BT008725.1 - 4 - 5 522 94

View Full Product Details