Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WCRKC2
Entrez Gene ID
830305
Gene Name
WCRKC thioredoxin 2
Gene Aliases
AT5G04260, T19N18.5, WCRKC thioredoxin 2
Chromosome Location
Chr.5: 1178644 - 1180060 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120508.4 NP_196046.2 1 - 2 348 90
GenBank mRNA AY063850.1 - 1 - 2 324 90
AY117290.1 - 1 - 2 310 90

View Full Product Details