Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDA
Entrez Gene ID
830315
Gene Name
limit dextrinase
Gene Aliases
AT5G04360, ATLDA, ATPU1, LIMIT DEXTRINASE, PU1, PULLULANASE, PULLULANASE 1, T19N18.90, T19N18_90, limit dextrinase
Chromosome Location
Chr.5: 1221448 - 1228841 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342767.1 NP_001330089.1 22 - 23 2376 72
NM_120518.5 NP_196056.2 22 - 23 2389 72

View Full Product Details