Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HY2
Entrez Gene ID
820069
Gene Name
phytochromobilin:ferredoxin oxidoreductase, chloroplast / phytochromobilin synthase (HY2)
Gene Aliases
ARABIDOPSIS ELONGATED HYPOCOTYL 2, AT3G09150, ATHY2, ELONGATED HYPOCOTYL 2, GENOMES UNCOUPLED 3, GUN3, PHYTOCHROMOBILIN SYNTHASE
Chromosome Location
Chr.3: 2803528 - 2805922 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337803.1 NP_001326171.1 2 - 3 461 109
NM_111750.4 NP_566344.2 2 - 3 476 109
NM_180665.1 NP_850996.1 2 - 3 401 109
NM_202535.1 NP_974264.1 1 - 2 577 109
GenBank mRNA AB045112.1 - 2 - 3 434 109
AY099706.1 - 2 - 3 477 109
AY128900.1 - 2 - 3 339 109

View Full Product Details