Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEL1
Entrez Gene ID
823971
Gene Name
DP-E2F-like 1
Gene Aliases
AT3G48160, DP-E2F-LIKE PROTEIN 1, DP-E2F-like 1, E2F-LIKE 3, E2FE, E2L3
Chromosome Location
Chr.3: 17783331 - 17786160 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114685.4 NP_190399.2 2 - 3 515 86
NM_180681.1 NP_851012.1 2 - 3 515 86
GenBank mRNA AB074533.1 - 2 - 3 205 86
AJ417836.1 - 2 - 3 212 86
BT004258.1 - 2 - 3 515 86
BT005524.1 - 2 - 3 205 86

View Full Product Details