Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUR5
Entrez Gene ID
824667
Gene Name
phosphoribosylformylglycinamidine cyclo-ligase, chloroplast / phosphoribosyl-aminoimidazole synthetase / AIR synthase (PUR5)
Gene Aliases
AT3G55010, ATPURM, EMB2818, EMBRYO DEFECTIVE 2818
Chromosome Location
Chr.3: 20386597 - 20388860 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115359.3 NP_191061.1 8 - 9 1089 73
NM_202708.2 NP_974437.1 7 - 8 1206 73
GenBank mRNA AY060585.1 - 8 - 9 1067 73
AY142054.1 - 7 - 8 993 73

View Full Product Details