Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl18
Entrez Gene ID
840248
Gene Name
serine carboxypeptidase-like 18
Gene Aliases
AT1G33540, F10C21.18, F10C21_18, serine carboxypeptidase-like 18
Chromosome Location
Chr.1: 12162112 - 12164714 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333051.1 NP_001319134.1 9 - 10 973 104
NM_001333052.1 NP_001321942.1 8 - 9 1052 104

View Full Product Details