Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATE2F2
Entrez Gene ID
841202
Gene Name
winged-helix DNA-binding transcription factor family protein
Gene Aliases
ARABIDOPSIS THALIANA HOMOLOG OF E2F C, AT1G47870, ATE2FC, E2FC, E2FC TRANSCRIPTION FACTOR, T2E6.2, T2E6_2, TRANSCRIPTION FACTOR-2 E2F2
Chromosome Location
Chr.1: 17634697 - 17637809 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198237.1 NP_001185166.1 3 - 4 656 96
NM_103684.3 NP_175222.1 3 - 4 719 96
GenBank mRNA AF242581.1 - 3 - 4 518 96
AJ417834.1 - 3 - 4 492 96
AY045636.1 - 3 - 4 579 96
AY058231.1 - 3 - 4 479 96

View Full Product Details