Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IPP1
- Entrez Gene ID
-
831505
- Gene Name
- isopentenyl diphosphate isomerase 1
- Gene Aliases
-
AT5G16440, ISOPENTENYL DIPHOSPHATE ISOMERASE, MQK4.17, MQK4_17, isopentenyl diphosphate isomerase 1
- Chromosome Location
-
Chr.5: 5371627 - 5373809 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121649.6 |
NP_197148.3 |
4 - 5 |
624 |
63 |
View Full Product Details