Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ER-ANT1
- Entrez Gene ID
-
831606
- Gene Name
- endoplasmic reticulum-adenine nucleotide transporter 1
- Gene Aliases
-
AT5G17400, T10B6.60, T10B6_60, endoplasmic reticulum-adenine nucleotide transporter 1
- Chromosome Location
-
Chr.5: 5728789 - 5730430 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121746.4 |
NP_568345.1 |
2 - 3 |
464 |
122 |
View Full Product Details