Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GLR5
- Entrez Gene ID
-
817801
- Gene Name
- glutamate receptor 5
- Gene Aliases
-
AT2G32400, ATGLR3.7, GLR3.7, GLUTAMATE RECEPTOR 3.7, T32F6.8, T32F6_8, glutamate receptor 5
- Chromosome Location
-
Chr.2: 13752263 - 13756893 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128799.4 |
NP_565744.1 |
5 - 6 |
2342 |
111 |
View Full Product Details